illustra™ PCR nucleotide mix dNTP set
Leverancier: Cytiva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
28-4065-64 28-4065-60
28-4065-64EA 110 EUR
28-4065-64 28-4065-60
illustra™ PCR nucleotide mix dNTP set
Reagentia voor nucleïnezuren Nucleotiden
PCR nucleotide mix dNTP set (2 mM, 10 mM, or 25 mM each A,C,G,T) for amplification, dideoxy sequencing, labelling, mutagenesis, cDNA synthesis, and expression profiling.
  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer-free and ready to use solutions in multiple formats
  • Functionally tested for long PCR and sequencing
Order Now
Specificaties
Environmentally Preferable
Applicatie