Extractors for specific light solvents
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05432037 05432049 05432058
LENZ05432037EA 462 EUR
LENZ05432037 LENZ05432049 LENZ05432058
Extractors for specific light solvents
Extractors Vloeistof-vloeistofextractors
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific light solvents.