Extractors for specific heavy solvents
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05433037 05433049 05433058
LENZ05433037EA 396 EUR
LENZ05433037 LENZ05433049 LENZ05433058
Extractors for specific heavy solvents
Extractors Vloeistof-vloeistofextractors
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific heavy solvents.