Extractors heads for specific heavy solvents
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
05433158 05433137 05433149
533-2152EA 332 EUR
533-2152 LENZ05433137 LENZ05433149
Extractors heads for specific heavy solvents
Extractors Vloeistof-vloeistofextractors
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific heavy solvents, with insert filter (porosity: 0).