Adapters, joint fiitings for reactor lids
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PC711.2438 PC711.1413 PC711.1913 PC711.1919 PC711.2413 PC711.2419 PC711.2913 PC711.2919 PC711.2925 PC711.2938 PC711.2425 PC711.3425 PC711.3419 PC711.3438 PC711.4519 PC711.4525 PC711.4538
COWIPC711.2438EA 32.1 EUR
COWIPC711.2438 COWIPC711.1413 COWIPC711.1913 COWIPC711.1919 COWIPC711.2413 COWIPC711.2419 COWIPC711.2913 COWIPC711.2919 COWIPC711.2925 COWIPC711.2938 COWIPC711.2425 COWIPC711.3425 COWIPC711.3419 COWIPC711.3438 COWIPC711.4519 COWIPC711.4525 COWIPC711.4538
Adapters, joint fiitings for reactor lids
Adapters Adapters, Slijpstukken met slangaansluitingen
PTFE, white.
  • Chemically inert
  • Reduce contamination and simplify cleaning
  • Non-scratch
Standard taper joint fittings for use with COWIE® PTFE reactor lids. Suitable for ‘A’ and ‘B’ length jointware. To assemble simply screw the tapered NPT joint into a corresponding NPT port of the reactor lid.