Spore strips, Attest™
Leverancier: 3M MEDICAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M1262 M1264
MMMEM1262EA 585 EUR
MMMEM1262 MMMEM1264
Spore strips, Attest™
Sterilisatie-indicatoren
3M™ Attest™ Biological Monitoring Systems are true biological indicators, in self-contained indicator capsules. They enable checks on the biological reliability and performance of each steriliser without laboratory facilities and laboratory trained personnel.
  • Self-contained, reducing the risk of false positives due to contamination
  • Colour change gives clear unambiguous, visual indication of PASS or FAIL
  • Time saving and results in 24 to 48 hours which require no interpretation