Soil hydrometers
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3470FC03868QP 3470FC06068QP 3470FC06068EU
614-1534EA 62.6 EUR
614-1534 ALLA3470FC06068QP ALLA3470FC06068EU
Soil hydrometers
Hydrometers
Soil hydrometers for specific gravity (SG) measurements.
  • Medium surface tension (55 mN/m)
  • Reading below meniscus
Certificaten: ASTM E100 D422 - A2LA traceable.
Bestelinformatie: EU traceable calibration certificate or EO official calibration certificate available on request.