Hydrometers
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3006TB070/60QP 3022FG080/60QP 3052TB065/60QP 3052FG150/60QP
ALLA3006TB070/60QPEA 34.1 EUR
ALLA3006TB070/60QP 614-1642 ALLA3052TB065/60QP ALLA3052FG150/60QP
Hydrometers
Hydrometers
Glass hydrometer calibrated at 60 °F (15,56 °C).
  • Graduated in specific gravity (SG)
  • Reading below meniscus
Verpakking: In a plastic case.