Hydrometers, Brix
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3910FC000/20QP 3910FC030/20QP 3910FC060/20QP
ALLA3910FC000/20QPEA 37.3 EUR
ALLA3910FC000/20QP ALLA3910FC030/20QP ALLA3910FC060/20QP
Hydrometers, Brix
Hydrometers
These Brix sugar scale hydrometers are specially designed for use in breweries.