Twaddle hydrometers
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
3930FG011/60QP 3930FG012/60QP 3930FG013/60QP 3930FG014/60QP
614-1622EA 22.7 EUR
614-1622 614-1623 614-1624 614-1625
Twaddle hydrometers
Hydrometers
Hydrometers for sugar with scales.
Calibration temperature of 15.6 °C.