Adhessive wound dressings, Tegaderm™
Leverancier: 3M MEDICAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
M1628 M1626W M1633 M1624W
MMMEM1628EA 225 EUR
MMMEM1628 MMMEM1626W MMMEM1633 MMMEM1624W
Adhessive wound dressings, Tegaderm™
Eerste-hulpbenodigdheden Wondverbanden
This transparent film dressing in frame style provides a waterproof, sterile barrier to external contaminants including liquids, bacteria and viruses. In vitro testing shows that the transparent film provides a viral barrier from viruses 27 nm in diameter or larger while the dressing remains intact without leakage.
  • Made with a hypoallergenic, latex-free adhesive that is gentle to the skin yet holds catheters and other devices in place
  • Conformable: dressing flexes with skin for greater patient comfort
  • Picture-frame delivery makes placement accurate and easy, minimising potential to stick to gloves or to itself