Dishes and lids
Leverancier: NICKEL ELECTRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
603 605 640 644
NICK603EA 0 EUR
NICK603 NICK605 NICK640 NICK644
Dishes and lids
Reservoirs/Schalen/Potten Laboratoriumbakken/-schalen
Made of aluminium.
  • Very good temperature resistance
  • Good chemical resistance
  • Lightweight