Conductivity electrodes
Leverancier: CONSORT
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SK21T SK10B SK10T SK24T SK23T SK20B SK20T
CONSSK21TEA 328 EUR
CONSSK21T CONSSK10B CONSSK10T CONSSK24T CONSSK23T CONSSK20B CONSSK20T
Conductivity electrodes
Elektroden Geleidbaarheidselektroden
A range of conductivity electrodes for different applications.
  • Available with or without integrated temperature sensor
  • Shaft is made from glass or epoxy