Gas cylinder depots, for storing gas cylinders outdoor
Leverancier: DUPERTHAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70-201340-001 70-201340-003 70-201040-001 70-200740-003 70-201040-003 70-200740-001
DUEH70-201340-001EA 1510 EUR
DUEH70-201340-001 DUEH70-201340-003 DUEH70-201040-001 DUEH70-200740-003 DUEH70-201040-003 DUEH70-200740-001
Gas cylinder depots, for storing gas cylinders outdoor
Meubilair Opslagkasten
Gas cylinder depots are used for storing gas cylinders outdoors in accordance with the regulations.
  • Robust construction made of sheet steel, material thickness 1,5 mm
  • Three-point locking system with door handle with security cylinder lock at an ergonomically favourable height
  • Two horizontal installation rails for mounting fittings
  • Prepared for ground fixing and earthing possibility on the ceiling
  • Natural ventilation and labelling with warning symbol W19
These gas cylinder depots have good corrosion resistance with polyester powder coating RAL 9002 grey white on galvanised base material.
Chequer plate floor (3 mm), including corrugation (4,5 mm).
Leveringsinformatie: Supplied with one cylinder holder with one retaining belt per standing space.