Drum pumps, mains operated
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5700-4400 5700-2200
BURK5700-4400EA 1270 EUR
BURK5700-4400 BURK5700-2200
Drum pumps, mains operated
Pompen Vatpompen
These are light- and high-performance pump to transfer small quantities of watery media up to 300 mPas from canisters, barrels and narrow necked containers from 32 mm opening diameter. Laboratory pumps, with splash water protected motor 230 V/200 W (IP 44), with 1,5 m PVC-hose, PP nozzle, 2 hose clips, discharge spout, wall bracket.
  • Made up of PP
  • Built-in safety key as on/off switch
  • Seal-less pump mechanism for acids and alkalis
  • Thread connection 1" with hose connector 3/4"