Absorbent skimmer socks, PIG®
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
104PS SKM203
NEWP104PSEA 110 EUR
NEWP104PS NEWPSKM203
Absorbent skimmer socks, PIG®
Absorptiematerialen en bescherming tegen morsen
Skimmer socks are used to absorb oil from acid baths. Chemical-resistant PP skin and filler pulls oils away from acids, chemicals, rainwater and other water-based liquids.
  • Easy to retrieve — floats on surface even after fully saturated
  • Skin remains UV resistant for up to 12 months
  • Attached string allows socks to be tied down, tied together or dipped into tanks
Certificaten: Meets NFPA 99 standards.