Drum pumps for acids and alkalis
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5762-0101 5762-0100
BURK5762-0101EA 2070 EUR
BURK5762-0101 BURK5762-0100
Drum pumps for acids and alkalis
Pompen Vatpompen
These drum pumps are suitable for weaker acids such as hydrochloric acid, phosphoric acid, chromic acid, battery acid, citric acid and more.
Leveringsinformatie: Supplied with pump tube, PP hose connection, PVC-fabric hose, PP nozzle, PP drum adapter.
Order Now
Specificaties
Environmentally Preferable