Jerricans, polyethlene
Leverancier: DUPERTHAL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5.09.010 5.11.010
DUEH5.09.010EA 30.7 EUR
DUEH5.09.010 DUEH5.11.010
Jerricans, polyethlene
Flessen Jerrycans
PE, electrostatically conductive.
  • Good chemical resistance
  • Good temperature resistance
  • Recyclable