PIG® SUPER absorbent socks
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PIG210 PIG211 PIG214
121-0537EA 125 EUR
121-0537 NEWPPIG211 NEWPPIG214
PIG® SUPER absorbent socks
Absorptiematerialen en bescherming tegen morsen
PIG® SUPER Absorbent Sock is great for high-volume machine leaks. Soaks up nearly twice as much liquid as other maintenance socks.
  • Extra-thirsty, formable socks hug corners and walls, surround higher-volume machine leaks and contain spills to keep workers safe
  • Cellulose filler is highly absorbent for big leaks and spills; super absorbency means that you replace them less often
  • Stitch-bonded, polypropylene skin resists bursting; reduces dust and holds in liquid, even when saturated
  • Absorbs and retains oils, coolants, solvents and water; not recommended for acids, caustics or other corrosive liquids
  • 85% recycled cellulose filler is ideal when green products are desired
Wringable for reclaiming up to 65% of absorbed liquid. Can be incinerated after use to reduce waste or for fuels blending.