Spaghetti booms, PIG®
Leverancier: NEW PIG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BOM202 BOM203 BOM204 BOM402 BOM403
NEWPBOM202EA 480 EUR
NEWPBOM202 NEWPBOM203 NEWPBOM204 NEWPBOM402 NEWPBOM403
Spaghetti booms, PIG®
Absorptiematerialen en bescherming tegen morsen
Surround and clean up thick oil spills on water with floating PIG® Spaghetti Booms.
  • Polypropylene filler is chemical-resistan and absorbs and retains oils and oil-based liquids—including lubricants, fuels and cleaning agents—without absorbing a drop of water
  • Connectors and rope running along length of boom facilitates extension, deployment and retrieval of boom(s)
  • Spaghetti strip design is ideal for pulling in thick liquids and for use in fast-moving water
  • Tough, bright white, outer mesh is UV-resistant for long-term outdoor use without degradation and