OpenLAB CDS EZChrom edition
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
906-0145EA 8330 EUR
906-0145 906-0146 906-0147 906-0148 906-0093 906-0097
OpenLAB CDS EZChrom edition
Software Chromatography Software
Scalable, compliant chromatography software platform with multi-instrument control. OpenLAB CDS EZChrom Edition is part of the Agilent software suite OpenLAB for analytical laboratories and enterprise. Its multi-instrument control capabilities, scalability and compliance are ideal to fulfil the requirements of today's chromatography laboratories, and to optimise their efficiency. Central administration functions have been arranged in the OpenLAB Shared Services/Control Panel module while keeping the existing user-interface and workflows for the EZChrom Elite™ chromatography data system. ManyMore Product Information