Capillary columns, BP5MS
Leverancier: Trajan Scientific and Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
054301 054310 054320 054321 054308
568-0328EA 638 EUR
568-0328 568-0327 SGEA054320 SGEA054321 568-0326
Capillary columns, BP5MS
Kolommen voor chromatografie
Optimised silphenylene content for general purpose MS analyses. Perfect for 5% MS analysis. Non polar, 5% equivalence. Low bleed, great inertness.
5% phenyl polysilphenylene-siloxane
Max. temperature: -40 to +330/350 °C
Suitable replacement for: DB-5ms, Rtx-5ms, HP-5MS, Rxt-Sil 5MS, AT-5ms, VF-5ms