Spray bottles with pump vaporiser, LaboPlast®
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0309-3025 0309-3005 0309-3100 0309-3007
215-3093EA 2.3 EUR
215-3093 215-3091 215-3092 BURK0309-3007
Spray bottles with pump vaporiser, LaboPlast®
Flessen Flessen met verstuiver
Bottle made of HDPE, spray pump made of PP and stainless steel.
  • Good chemical resistance
  • Good temperature resistance
Spray bottles with pump vapouriser and safety cap for the pump. Liquids can be finely vapourised at the push of a button in an environmentally compatible way. The transparent bottles make identification of contents easy. The protective cover prevents unintentional pressing of the pump and acts as a dust cover.