Organic size exclusion chromatography columns, GPC KD-800 series
Leverancier: Resonac
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F6028290 F6028210 F6028220 F6028225 F6028230 F6028240 F6028250 F6028260 F6028270 F6700411
554-2844EA 1822 EUR
554-2844 554-2836 554-2837 554-2838 554-2839 554-2840 554-2841 554-2842 554-2843 554-2882
Organic size exclusion chromatography columns, GPC KD-800 series
Kolommen voor chromatografie
Standard organic solvent SEC (GPC) column.
  • Supports a wide range of applications from low to high molecular weight compounds
* Estimated value