Calibration weights, OIML, class M2
Leverancier: Sartorius Balances
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
YCW556-AC-00 YCW626-AC-02 YCW7169-AC-00 YCW616-AC-02 YCW626-AC-00
SARBYCW556-AC-00EA 63.7 EUR
SARBYCW556-AC-00 SARBYCW626-AC-02 SARBYCW7169-AC-00 SARBYCW616-AC-02 SARBYCW626-AC-00
Calibration weights, OIML, class M2
Gewichten Kalibratiegewichten
Weights are used for control of inspection, measuring and test equipment.
Proof Line series, OIML Class M2 knob weights are manufactured from brass, precision-lathed, fine finish with the density of 8,400 kg/m3 and the block weights are made up of cast iron, gray cast, black lacquered finish with the density of 7,200 kg/m3.
Certificaten: Compliance with OIML R111-1.