Precision thermometers, Safe BIO-TEMP®
Leverancier: Alla France
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5980T020-T-QP 5980T030-T-QP 5980T050-T-QP
ALLA5980T020-T-QPEA 124 EUR
ALLA5980T020-T-QP ALLA5980T030-T-QP ALLA5980T050-T-QP
Precision thermometers, Safe BIO-TEMP®
Thermometers Analoge thermometers
Mercury-free thermometers with yellow back glass.
  • PTFE coated thermometer filled with environmentally safe blue organic filling liquid, for extra security
  • A high density PE base houses the reservoir of the thermometer, acting as a type of thermal brake
  • Instrument can be free-standing or attached
Bestelinformatie: Supplied with adhesive strip and magnet, in a recyclable sturdy plastic concertina box, ideal for secure storage when not in use, and with a conformity certificate, (on request) can be supplied with 'EU' Traceable calibration certificate.