Reversed phase chromatography columns, silica-based C18M
Leverancier: Resonac
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F6650041 F6650040
SHOWF6650041EA 711 EUR
SHOWF6650041 SHOWF6650040
Reversed phase chromatography columns, silica-based C18M
Kolommen voor chromatografie
C18M are monometric type ODS columns, fully endcapped high purity silica (99,99% or higher). Ideal for reversed phase separation.