PIG® fibreglass chemical trays
Leverancier: New Pig
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PIG®
NEWPPAK743-BKEA 0 EUR
NEWPPAK743-BK NEWPPAK739-WH NEWPPAK742-WH NEWPPAK743-WH NEWPPAK744-WH NEWPPAK740-WH NEWPPAK741-WH
PIG® fibreglass chemical trays
Reservoirs/Schalen/Potten Trays/pannen voor gemorste stoffen
Catch drips and control messes with these smooth, seamless fibreglass trays.
  • Reinforced thermoset composite fibreglass resists chemicals and retains its shape, even under heavy loads
  • Smooth corners and seamless design allows trays to be steam cleaned or sterilized for repeated use
  • Molded edges safely contain minor spills or leaks from containers or oily parts
Chemical-resistant trays are easy to clean.