Reversed phase chromatography columns, ODP2 HP
Leverancier: Resonac
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
F6714010 F6714011 F7622004 F7622005 F7622002 F7622001 F7622003
554-2903EA 456 EUR
554-2903 554-2904 554-2946 554-2947 554-2944 554-2943 554-2945
Reversed phase chromatography columns, ODP2 HP
Kolommen voor chromatografie
Provides nearly twice as large theoretical plate number compared with generally available polymer-based reversed phase columns.
  • Column offers enhanced retention of high polar substances compared with ODS column
  • Suitable for reversed phase analysis of small molecules such as pharmaceutical compounds in the presence of protein matrix
  • Ideal for LC-MS analysis of high polar compounds