Racks for chest freezers
Leverancier: GLW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E318 E321 K120
GLWAE318EA 206 EUR
GLWAE318 GLWAE321 GLWAK120
Racks for chest freezers
Rekken Rekken voor cryogene dozen
These freezer rack are commonly used for all chest freezers and available in various exterior heights.
  • Made of stainless steel
  • Can accomodate 18 boxes
  • With one folding handle on top
Bestelinformatie: Stainless steel locking rod has to be ordered separately.