Laryngeal mirrors
Leverancier: KLS Martin Group
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
39-781-10 39-781-22 39-781-30
MART39-781-10EA 0 EUR
MART39-781-10 MART39-781-22 MART39-781-30
Laryngeal mirrors
Spiegels
Laryngeal mirrors are used for tonsillectomy and laryngo-bronchoscopy.
  • Made up of stainless steel 18/8
  • Antimagnetic
  • Acid resistant