Sleeve Sampler
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1035A-1000 1035T-150 1035E-1000 1035T-200 1035P-150 1035A-1500
SAMP1035A-1000EA 457 EUR
SAMP1035A-1000 SAMP1035T-150 SAMP1035E-1000 SAMP1035T-200 SAMP1035P-150 SAMP1035A-1500
Sleeve Sampler
Monsternemers Monsternemers voor vaste stoffen
The sleeve sampler is specially designed for large volumes at great depths.
  • Adjustable length and bright finish design
  • Extremely strong and robust extension rods
Suitable for free-flowing powders and granules.