Slot samplers
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1210A-600 1210A-1000 1210A-1500 1210B-1000 1210K-1000 1210A-300 1210A-2000 1210K-1500 1210K-2000
SAMP1210A-600EA 774 EUR
SAMP1210A-600 SAMP1210A-1000 SAMP1210A-1500 SAMP1210B-1000 SAMP1210K-1000 SAMP1210A-300 SAMP1210A-2000 SAMP1210K-1500 SAMP1210K-2000
Slot samplers
Monsternemers Monsternemers voor vaste stoffen
These all layer samplers have a robust all stainless steel construction and are suitable for sampling free flowing powders, granules and even slightly cohesive powders.
  • Multilevel samplers
  • Cross section type
  • With or without bottle holder
For taking a large volume, cross sectional sample from a container.
Bestelinformatie: Sample collecting bottles (for use with the sampler with bottle holder) as well as cleaning brushes can be ordered separately.