Slot samplers, SteriWare®
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8073H-1000 8073H-500 8073H-1000S 8073H-500S 8073J-1000S 8073J-1000 8073H-300
300-0041EA 360 EUR
300-0041 300-0043 300-0042 300-0044 300-0217 SAMP8073J-1000 SAMP8073H-300
Slot samplers, SteriWare®
Monsternemers Monsternemers voor eenmalig gebruik
These cross sectional multilevel disposable powder samplers are designed for powders. The robust HDPE construction make this to be used in huge number of testing applications.
  • BSE/TSE-free
  • Full batch traceability
  • Suitable for use in ATEX zones 0, 1 and 2
  • Available as gamma irradiated sterile (SAL 10-6)
Can take a composite from the top, middle and bottom of the powder bed – Moulded and packed in a cleanroom to ensure cleanliness.
Ideal for taking a multilevel, cross sectional sample from powders and granules. Insert the sampler and rotate the handle to take a representative sample.
Certificaten: FDA, EU 10/2011 and EC 1935/2004.
Verpakking: Individually bagged in a heat-sealed PE bag.