Adapter, stopper type, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286576503 286576803 286576109 286576306 286576709 286576906
SCOT286576503EA 376 EUR
SCOT286576503 SCOT286576803 SCOT286576109 SCOT286576306 SCOT286576709 SCOT286576906
Adapter, stopper type, DURAN®
Adapters Adapters met NS-verbindingen
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Adapters, stopper type, solid, socket and cone.