Adapters, expansion, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286542507 286542704 286548004 286548107 286548201 286535906
SCOT286542507EA 202 EUR
SCOT286542507 SCOT286542704 SCOT286548004 SCOT286548107 SCOT286548201 SCOT286535906
Adapters, expansion, DURAN®
Adapters Adapters met NS-verbindingen
DURAN®, borosilicate glass 3,3 clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapter from small cone to large socket.