Powder samplers
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1220A-1500 1220A-600 1220A-900 1220B-600 1220B-900 1220B-1500 1220B-300 1220J-900 1220J-1500 1220J-2000
SAMP1220A-1500EA 1150 EUR
SAMP1220A-1500 SAMP1220A-600 SAMP1220A-900 SAMP1220B-600 SAMP1220B-900 SAMP1220B-1500 SAMP1220B-300 SAMP1220J-900 SAMP1220J-1500 SAMP1220J-2000
Powder samplers
Monsternemers Monsternemers voor vaste stoffen
Pocket samplers for discrete zone samples from powders and granules.
  • Simple operation and robust design
  • Stainless steel or PTFE inner rod
  • Can be stripped and cleaned very quickly
Suitable for use in pharmacy applications.
Leveringsinformatie: Supplied with pointed tips as standard.