Gaswasflessen, volgens Drechsel, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
247142404 247143606 247144405
SCOT247142404EA 34.8 EUR
SCOT247142404 SCOT247143606 SCOT247144405
Gaswasflessen, volgens Drechsel, DURAN®
Flessen Gaswasflessen
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Bottle without head.