Sleeve samplers, U-D Master
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1500A-01 1500A-02 1500A-03 1500A-04 1500S-01 1500W-01 1500W-02 1500W-04 1500W-12 1500W-17 1500W-18 1500W-22 1500W-08 1500W-03
SAMP1500A-01EA 270 EUR
SAMP1500A-01 SAMP1500A-02 SAMP1500A-03 SAMP1500A-04 SAMP1500S-01 SAMP1500W-01 SAMP1500W-02 SAMP1500W-04 SAMP1500W-12 SAMP1500W-17 SAMP1500W-18 SAMP1500W-22 SAMP1500W-08 SAMP1500W-03
Sleeve samplers, U-D Master
Monsternemers Monsternemers voor vaste stoffen
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
  • Quick and easy accurate unit dose sampling
  • Different volumes can be taken using the same sampler
  • Simply change the tip for a different volume
Designed for use with free flowing powders and granules.