Roerhulzen, KPG®, DURAN®
Leverancier: DWK Life Sciences
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
286303803 286304002 286304208 286304405 286304602 286304808
SCOT286303803EA 260 EUR
SCOT286303803 SCOT286304002 SCOT286304208 SCOT286304405 SCOT286304602 SCOT286304808
Roerhulzen, KPG®, DURAN®
Roerders Roergereedschap
DURAN®, clear borosilicate glass 3.3
DURAN® KPG® bearing sleeves are used as components for agitator guides/agitator coolers. They are available as ground or polished versions in the following designs:
HA: without grease cup
HB: with grease cup