Tubings, braided, for cooling water
Leverancier: HUBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
16853 16851 16852 16854 16855 16856 16857 16858 16859
HUBE16853EA 92 EUR
HUBE16853 HUBE16851 HUBE16852 HUBE16854 HUBE16855 HUBE16856 HUBE16857 HUBE16858 HUBE16859
Tubings, braided, for cooling water
Slangen Transfer Tubing
Flexible braided hoses suitable for water and water/mono ethylene glycol mixtures up to 50%.