Flash chromatography columns, PuriFlash®, C18 IR silica
Leverancier: INTERCHIM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IR50C18/450G IR-50C18-F0025 IR50C18/35G IR-50C18-F0330 IR50C18/2500G IR50C18/20G IR50C18/175G IR50C18/1250G IR50C18/6G
INTEIR50C18/450GEA 0 EUR
INTEIR50C18/450G UPTIIR-50C18-F0025 INTEIR50C18/35G UPTIIR-50C18-F0330 INTEIR50C18/2500G INTEIR50C18/20G INTEIR50C18/175G INTEIR50C18/1250G INTEIR50C18/6G
Flash chromatography columns, PuriFlash®, C18 IR silica
Kolommen voor chromatografie
These flash chromatography columns, irregular C18 silica serves a broad-ship of purification requirements for mid to non polar compounds.
  • Base material is irregular shaped silica
  • pH ranges from 1,6 to 7