Lockings for upright/chest freezer racks
Leverancier: GLW
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
V1 V14 V19 V24 V30 V36 V42 V48 V52 V58 V62 V65 V68
GLWAV1EA 4.4 EUR
GLWAV1 GLWAV14 GLWAV19 GLWAV24 GLWAV30 GLWAV36 GLWAV42 GLWAV48 GLWAV52 GLWAV58 GLWAV62 GLWAV65 GLWAV68
Lockings for upright/chest freezer racks
Veiligheidssloten
These stainless steel locking bracket and rods for freezer racks can be used to secure the individual trays in the racks.
They are compatible with all GLW-E-series stainless steel racks (locking bracket not compatible with E318 and E321).