Disposable weighing boats, Sterilin®
Leverancier: STERLIN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
WB30328 WB30322
SLINWB30328EA 272 EUR
SLINWB30328 SLINWB30322
Disposable weighing boats, Sterilin®
Schuitjes Weegschuitjes
Square shaped PS weighing boats. Raw material modified to significantly reduce the charge relaxation time in comparison to standard weigh boats. Majority of electrostatic charge dissipates within seconds when earthed on the balance pan.
  • Avoids the retention of powders by static electricity
  • Ideal for use with fine powders e.g. talcum powder