LabSafe™ GEL blue for staining of polyacrylamide gels
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-35 786-35G
786-35EA 437 EUR
786-35 786-35G
LabSafe™ GEL blue for staining of polyacrylamide gels
Kleurstoffen Kleuringen voor elektroforese
LabSafe GEL Blue™ is an enhanced protein stain based on Coomassie dye that offers unsurpassed sensitivity and rapid band visualisation.
  • Sensitive, 4-8ng BSA
  • Saves time, band develops within 3 minutes
  • Avoids the use of methanol, acetic acid, or other toxic agents
  • Works with native PAGE, SDS-PAGE, isoelectric focusing, or 2D gels
LabSafe GEL Blue™ is an ultra sensitive stain that is able to detect as little as 4ng protein. Protein bands are visible in as little as 3 minutes, with maximal staining achieved in an hour. The figure shows that LabSafe GEL Blue™ can detect protein levels as low as 4ng BSA. 60 to 1000ng BSA is detectable in 5 to 10 minutes, and the low levels of BSA (<8ng) become clearly visible when washed in water.
Works with native PAGE, SDS-PAGE, denaturing, isoelectric focusing, or 2D gel. Mass spectrometry compatible.