Silver ≥99.9% (metals basis), nanopowder APS 20-40 nm
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
45509.06 45509.14
45509.06EA 240 EUR
45509.06 45509.14
Silver ≥99.9% (metals basis), nanopowder APS 20-40 nm
Silver
Purity: 99.9% CAS No.: 7440-22-4 Molecular Formula: Ag Physical Form: No Oxide layer Odour: Characteristic UN No.: 3077 MDL No.: MFCD00003397
Formule: Ag
MW: 107.87 g/mol
Kookpunt: 2212
Smeltpunt: 962
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
EINECS: 232-033-3
UN: 3077
ADR: 9 III
Merck Index: 12,08647