ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Leverancier: Serva
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30315.02 30315.03 30315.04 30315.05
SERA30315.02EA 35 EUR
SERA30315.02 SERA30315.03 SERA30315.04 SERA30315.05
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na2, oxidized form)
Formule: C₂₁H₂₆N₇Na₂O₁₇P₃
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00065390
CAS nummer: 24292-60-2
EINECS: 246-129-8