β-Nicotinamide adenine dinucleotide phosphate (β-NADP)
Leverancier: Serva
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
NADP+, Nicotinamide adenine dinucleotide phosphate, oxidized, Coenzyme II, β-Triphosphopyridine Nucleotide, TPN, Nadide phosphate, 1-((2R,3R,4S,5R)-5-((((((((2R,3R,4R,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxy-4-(phosphonooxy)tetrahydrofuran-2-yl)methoxy)(hydroxy)phosphoryl)oxy)oxidophosphoryl)oxy)methyl)-3,4-dihydroxytetrahydrofuran-2-yl)-3-carbamoylpyridin-1-ium
30313.01
30313.02
SERA30313.01EA
0
EUR
SERA30313.01
SERA30313.02
β-Nicotinamide adenine dinucleotide phosphate (β-NADP)
ß-Nicotinamide adenine dinucleotide phosphate (β-NADP)
Formule:
C₂₁H₂₈N₇O₁₇P₃ MW: 743.41 g/mol |
MDL Nummer:
MFCD00067537 CAS nummer: 53-59-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...