Cyclopentylacetic acid 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2-Cyclopentylacetic acid

154870100 154870250
ACRO154870100EA 77.3 EUR
ACRO154870100 ACRO154870250
Cyclopentylacetic acid 97%
Cyclopentylacetic acid
Formule: C₇H₁₂O₂
MW: 128.17 g/mol
Kookpunt: 133…134 °C (23 mmHg)
Dichtheid: 1.02 g/cm³
Vlampunt: 109 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00001387
CAS nummer: 1123-00-8
EINECS: 214-368-7

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4520 to 1.4540 (20°C, 589 nm)
Titration with NaOH ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR