L-(-)-tert-Leucine (H-Tle-OH)

Leverancier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: (S)-2-Amino-3,3-dimethylbutanoic acid, H-Tle-OH

4006523.0001 4006523.0005
BACE4006523.0001EA 0 EUR
BACE4006523.0001 BACE4006523.0005
L-(-)-tert-Leucine (H-Tle-OH)
L-(-)-tert-Leucine (H-Tle-OH)
Formule: C₆H₁₃NO₂
MW: 131.2 g/mol
MDL Nummer: MFCD00064218
CAS nummer: 20859-02-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR